Learn More
STEMCELL Technologies RoboSep™ Mouse CD138 Positive Selection Kit, For processing: 2 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 18957RF
Encompass
The EasySep™ Mouse CD138 Positive Selection Kit is designed to isolate CD138+ cells from mouse splenocytes, lymph nodes and bone marrow by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell culture, and hybridoma generation.
Catalog No. 50-197-4661
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.