Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Chemistry Systems Solutions and Accessories

Catalog No. AVLBP0521 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVLBP0521 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AVLBP0521 Supplier Roche Diagnostics Supplier No. 03110435180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Type DeProteinizer Solution
For Use With (Application) To remove proteins from electrodes
Quantity 125 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.