Learn More
Stoelting™ Rochester-Pean Hemostatic Forceps
SDP
Catalog No. 10001346 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
5.5 in.
6.25 in.
7.25 in.
10 in.
Model:
Curved
Straight
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-001-346 6.25 in. Straight
10-001-344 5.5 in. Straight
10-001-345 5.5 in. Curved
10-001-347 6.25 in. Curved
10-001-348 7.25 in. Straight
10-001-350 7.25 in. Curved
10-001-351 10 in. Straight
10-001-352 10 in. Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 10-001-346 Supplier Stoelting™ Supplier No. 5211028
Add to Cart
Add to Cart
Stoelting™ Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
Specifications
For Use With (Application) Neuroscience
Length (English) 6.25 in.
Model Straight