Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Rock and Mineral Collection
SDPSCI_ED

Catalog No. S89547 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89547 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89547 Supplier United Scientific™ Supplier No. UNROCMIN20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

United Scientific™ Rock and Mineral Collection has 20 Types of Specimens (10 Rocks Specimens of Igneous, Sedimentary, and Metamorphic Rocks Types; 10 Minerals Specimens).

  • United Scientific Rock and Mineral Collection has 20 Types of Specimens, (10 Rocks Specimens of Igneous, Sedimentary and Metamorphic Rocks Types and 10 Minerals Specimens).
    • Rock Specimens include Basalt, Granite, Pumic, Gniess, Marble, Quartizite, Serpentine, Bituminous Coal, Limestone, Sand Stone.
    • The Minerals Specimens include Agate, Calcite, Dolomite, Feldspar, Graphite, Gypsum, Magnetite, Pyrophilite, Quartz, Tourmaline
    • Average sizes are 2 x 3 x 1.5 cm

Specifications

Grade 5 to 12
Format Kit
Product Type Rocks and Minerals
Includes 20 Specimens
Languages English
Subject Geology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.