Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rockin' Math Game
SCI_ED

Catalog No. S04110 Shop All Educational Insights Products
Change view
Click to view available options
Grade:
3 to 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04110 3 to 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04110 Supplier Educational Insights Supplier No. EI3222

missing translation for 'validMainframeMsg'

Climb the stairway to math super stardom!

  • Rock your way around the board, answering amped-up math questions at the recording studio, Glitz Mall, release party and benefit concert
  • Answer questions like: How many concert tickets do you have to sell to cover your agent's fees? How much leather will you need to make custom pants for your entire band?

Specifications

Grade 3 to 4
Product Type Rockin' Math Game
Includes Guitar-shaped box, 220 pick-shaped cards in limo holder and 4 star-shaped playing pieces
For Use With (Application) Climb the stairway to math super stardom!
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.