Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) w Lecithin, Polysorbate 80 (LP80), Penicillinase

Catalog No. B215273 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
Packaging Type:
Sterile Double Wrapping Plate
Triple Wrapping Isolator Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B215273 100/Pk. Triple Wrapping Isolator Pack
B215274 100/Pk. Sterile Double Wrapping Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B215273 Supplier BD Supplier No. 215273
Add to Cart
Add to Cart
This item is not returnable. View return policy

The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.

The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:

  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Country of Origin United States
Additive Lecithin, Polysorbate 80
Form Liquid
Format Plate, Sterile, Isolator Pack
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with Lecithin and Polysorbate 80 and Penicillinase
Sterility Sterile
For Use With (Application) Rodac™ snap lid has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate
Packaging Type Triple Wrapping Isolator Pack
Quantity 100/Pk.
Protein Source Casein, Soy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.