Description
- Suitable for work performed under vacuum and pressure
- The connections remain absolutely stable and can be easily separated again due to components that are technically matched to each other
- The connection comprises cone and socket as well as cap, safeguarding and sealing ring
- Can be used up to a temperature of 200°C
- Minimum breakage risk due to anti-jamming system
- Replaceable with ground joints in accordance with DIN 12 249
Specifications
Specifications
| Connection Type | B14/23 |
| Type | Rodaviss Connecting Set |
| For Use With (Application) | Optimizing Glass Connections |
| For Use With (Equipment) | Carousel 6 Plus Reaction System |
| Quantity | 10/Pk. |
| Temperature (Metric) Range | Up to 200°C |