Learn More
Stoelting™ Rodent Ventilator
SDP
Catalog No. 10001071 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
1 mL
5 mL
10 mL
30 mL
Model:
Synchronized start/stop Ventilator
Ventilator
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Model
10-001-071 30 mL Ventilator
10-000-625 1 mL Ventilator
10-000-976 10 mL Synchronized start/stop Ventilator
10-000-977 5 mL Synchronized start/stop Ventilator
10-001-069 10 mL Ventilator
10-001-070 5 mL Ventilator
10-001-096 30 mL Synchronized start/stop Ventilator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-001-071 Supplier Stoelting™ Supplier No. 500953
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Rodent Ventilator features a unique design that ensures accuracy, durability and quietness of operation unequalled by any other ventilator.
  • Unique design, compact and reliable
  • Quiet operation
  • Negligible R.F. broadcasting
  • Ideal for use with mice and is also suitable for small birds and perinatal rats.
  • Valve mechanism features nearly zero dead space and superb durability with no leakage
  • Expired air may be channeled for analysis when used with anesthetic gases
  • Variable stroke linkage mechanism, very similar in principle to the one used in Basile's larger respirators, operates the piston
  • Reciprocating motion is adjusted and transmitted to the piston by rods and articulated joints only, which leads to minimal wear, no backlash, silent operation and exact stroke reproducibility
  • Stroke volume is adjustable whether the ventilator is on or off. For the most convenient and accurate stroke volume control available, the knob and scale remain stationary, even when the piston is in motion.
  • Stroke rate (breaths per minute) is feedback controlled from 50 to 300 strokes/min and displayed on an LED digital readout. A connection square of four ports (pictured at right) include intake, out to animal, return from animal and exhaust
  • Optional controlled pause feature available
  • Rat and mouse units are supplied with one of two easily interchangeable cylinder/piston assemblies that enable a broader range of stroke volumes.
Specifications
For Use With (Application) Neuroscience
Size 30 mL
Model Ventilator