Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ ROI & Background Calibration Kit, Fast 96-well

Catalog No. 50612238
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-612-238 2 x 96 well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-612-238 Supplier Applied Biosystems™ Supplier No. 4432638
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to maintain your real-time PCR instrument with Fast 96-well block.

Region of interest (ROI) and background calibration should be run after lamp change.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C ± 5°C.

Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Reaction Speed Fast
Product Type Background Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 2 x 96 well plates
Calibration Type Spectral Calibration
PCR Method qPCR
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.