Learn More
TARGETMOL CHEMICALS INC ROLIPRAM 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6644200MG
Encompass_Preferred
Also available in 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rolipram (SB 95952) is a phosphodiesterase 4 inhibitor with antidepressant properties. purity: 99%
Catalog No. 50-257-6481