Learn More
Description
- Highly visible orange signs with contrasting black messages and symbols
- Lightweight, flexible fabric
- Non-reflective PVC extruded over polyester strands
- Ideal for daytime only applications
- Designed with four corner pockets to insert cross-ribs (sold separately)
Specifications
Specifications
| For Use With (Equipment) | Cross-Ribs (Sold Separately) |
| Material | Vinyl Mesh |
| Mounting Type | Corner Holes |
| For Use With (Application) | Temporary Traffic Control (TTC) Construction Zones |
| Dimensions (L x W) | 36 x 36 in. |
| Displayed Text | DETOUR AHEAD |
| Product Type | Warning Sign |
| Color | Black on Orange |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.