Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Roll-Up Construction Sign

Catalog No. 19169604 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
BE PREPARED TO STOP
DETOUR AHEAD
Flagger Pictogram
STREET CLOSED AHEAD
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-169-604 Flagger Pictogram
19-169-605 BE PREPARED TO STOP
19-169-603 DETOUR AHEAD
19-169-606 STREET CLOSED AHEAD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-169-604 Supplier Accuform™ Supplier No. FRC303MV
Add to Cart
Add to Cart

Accuform™ Roll-Up Construction Signs, for Temporary Traffic Control (TTC) construction zones, are quick to set up and easy to take down for storage

  • Highly visible orange signs with contrasting black messages and symbols
  • Lightweight, flexible fabric
  • Non-reflective PVC extruded over polyester strands
  • Ideal for daytime only applications
  • Designed with four corner pockets to insert cross-ribs (sold separately)

Specifications

For Use With (Equipment) Cross-Ribs (Sold Separately)
Material Vinyl Mesh
Mounting Type Corner Holes
For Use With (Application) Temporary Traffic Control (TTC) Construction Zones
Languages English
Dimensions (L x W) 36 x 36 in.
Displayed Text Flagger Pictogram
Product Type Warning Sign
Color Black on Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.