Learn More
Essendant Direct Thermal Printing Thermal Paper Rolls, 3"" x 225 ft, White, 24/Carton
Supplier:  Essendant ICX90781294
Encompass_Preferred
The paper rolls are guaranteed to work efficiently with your machine printhead to provide crisp, clear, black images. The reliability and performance of a thermal printer is directly related to the quality of the supplies, including the paper, used by the customer. The IBM Testing Process is an "Industry Standard" test for the thermal papers used in the marketplace. To be IBM approved, the paper must pass a 100km of paper printhead test without abrasion or build-up, with no cutting failures. IBM testing only confirms that the use of approved papers will not negatively affect the operating life of an IBM thermal printer. Manufactured to OEM specifications.
Catalog No. 26-801-42
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.