Learn More
Medchemexpress LLC Rolofylline | 136199-02-5 | >98.0% | 356.46 | C20H28N4O2 | 5 MG

Supplier: Medchemexpress LLC HY109655MG
Rolofylline is a selective adenosine A1 receptor antagonist supplied for research and analytical use. The compound has formula C20H28N4O2 and a molecular weight of 356.46 g/mol. It is provided as a solid for laboratory studies of adenosine receptor pharmacology and renal/diuretic research. Recommended storage: powder at -20°C; in solvent, -80°C for long-term stability.
- Selective adenosine A1 receptor antagonist suitable for pharmacology studies.
- Provided as a solid analytical and research standard for laboratory use.
- Chemical formula C20H28N4O2; molecular weight 356.46 g/mol.
- Storage: powder -20°C; in solvent -80°C for long-term stability.
- Available in small masses for solution preparation (e.g., 1 mg, 5 mg, 10 mg).
- High purity reported by third-party suppliers (≥98% by HPLC).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.