Learn More
TARGETMOL CHEMICALS INC ROMIDEPSIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6006
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Romidepsin (NSC-630176) is an intravenously administered histone deacetylase inhibitor and antineoplastic agent that is approved for use in refractory or relapsed cutaneous and peripheral T cell lymphomas. purity: 99%
Catalog No. 50-257-9686
Only null left
Edge
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.