Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570392 Supplier Invitrogen™ Supplier No. PA570392
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

The protein encoded by this gene is a mitochondrial membrane protein that is responsible for increasing the level of reactive oxygen species (ROS) in cells. The protein also has antimicrobial activity against a variety of bacteria by inducing bacterial membrane breakage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROMO1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ROMO1
Gene Accession No. P60603
Gene Alias 2010100O12Rik; AI853864; bA353C18.2; C13H20ORF52; C20orf52; Epididymis tissue protein Li 175; glyrichin; hypothetical protein MGC73345; mitochondrial targeting GxxxG motif protein; mitochondrial targeting GXXXG protein; MTGM; MTGMP; PCM19; protein MGR2 homolog; RA_r333_jsm669B4f; Reactive oxygen species modulator 1; romo1; ROS modulator 1; Unknown (protein for MGC:127858); zgc:73345; zgc:77111
Gene Symbols ROMO1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ROMO1 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67067
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.