Learn More
Roosterbio Inc RoosterVial™-hBM: Human Bone Marrow-Derived Mesenchymal Stem/Stromal Cells
SDP
Supplier:  Roosterbio Inc MSC003
Encompass_Preferred
RoosterVial™-hBM are Human Bone-Marrow Derived Mesenchymal Stem/Stromal Cells (hBM-MSCs) isolated from adult human iliac crest from a variety of normal, healthy donors and expanded to a population doubling level (PDL) of 6-10, post-mononuclear cell isolation. Available in both 1M or high-volume 10M cell per vial formats to meet your needs throughout product development.Each lot is release tested to expand >10-fold within 4 to 6 days in a media exchange free process when paired with RoosterNourishTM-MSC bioprocess medium and conforms to ISCT minimal criteria for hMSC identity giving added confidence in quality, reproducibility, and ease of integration.Designed for robust, reproducible, standardized scale-up and supported by our media exchange free 2D and 3D bioreactor processes and RoosterNourish™-MSC expansion media, RoosterVial™-hBM can efficiently and reproducibly get you the millions or billions of hMSCs to accelerate your Research and Product Development.
Catalog No. NC2253325
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.