Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ropivacaine hydrochloride

Catalog No. AC467220010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467220010 1 g
AC467222500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467220010 Supplier Thermo Scientific Chemicals Supplier No. 467220010
Add to Cart

CAS: 98717-15-8 | C17H27ClN2O, C17H26N2O·HCl | 310.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98717-15-8
Molecular Formula C17H26N2O·HCl

Specifications

Quantity 1 g
Melting Point 149.0°C to 151.0°C
Formula Weight 328.88
InChI Key NDNSIBYYUOEUSV-RSAXXLAASA-N
SMILES Cl.CCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C
IUPAC Name (2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride
Molecular Weight (g/mol) 310.87
Chemical Name or Material Ropivacaine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.