Learn More
Description
Expected from sequence similarity: Bovine, Chimpanzee, canine and mouse. Immunogen sequence: KDRMEMPTAT QKTDTG & ARLDSDVSPL ETESYL.
Specifications
Specifications
| Antigen | ROPN1L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | Ropn1l |
| Gene Accession No. | Q96C74 |
| Gene Alias | Akapasp; AKAP-associated sperm protein; A-kinase anchoring protein-associated sperm protein; ASP; AV047578; radial spoke head 11 homolog; rhophilin associated tail protein 1 like; rhophilin associated tail protein 1-like; ROPN1L; ROPN1-like protein; ropporin 1-like; ropporin-1-like protein; RSPH11 |
| Gene Symbols | Ropn1l |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
