Learn More
Abcam Roquefortine C, Neuroto x in, 1MG
Supplier: Abcam AB1443451MG
MW 389.4 Da, Purity >98%. Mycotoxin. Potent neurotoxin. Inhibits Cytochrome p450. Inhibits lymphocyte proliferation. Penicillin (ab145634, ab143435) metabolite. Shows tremorgenic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.