Learn More
Selleck Chemical LLC Roquinimex S2307-5mg
SDP
Supplier:  Selleck Chemical LLC S23075MG
Encompass_Preferred
Roquinimex (FCF 89 Linomide LS 2616) is a novel immunomodulator augmenting natural killer (NK) cell activity and T-lymphocyte related effector functions Roquinimex is a potent oral active tumor antiangiogenic agent Roquinimex inhibits the secretion of TNF-
Catalog No. 50-311-1870
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.