Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ROR gamma Plasmid
SDP

Catalog No. NBP225278 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225278 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225278 Supplier R&D Systems™ Supplier No. NBP225278

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For expression of hROR gamma protein in mammalian cells, induction of IL-17A promoter activity

The ROR gamma Plasmid is a mammalian expression vector construct, pCMV/hRORgamma, containing coding sequence of human RAR-related orphan receptor gamma.

  • Species: Human
  • Gene: RORC
  • Immunogen: Human ROR gamma Plasmid
  • Total contents of 10μg is enough for one 96-well cell transfection

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Line Novus
Product Type ROR gamma Plasmid
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.