Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RORγt, Mouse Anti-Mouse, RY703, Clone: Q31-378, BD Horizon™

Catalog No. BDB571451 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg;
Conjugate:
RY703
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB571451 0.1 mg RY703
BDB571452 25 μg; RY703
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571451 Supplier BD Biosciences Supplier No. 571451
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RORγt Monoclonal antibody specifically detects RORγt in Mouse (QC Testing) samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RORγt
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone Q31-378
Concentration 0.2 mg/mL
Conjugate RY703
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias RORγT; RORgt; RORgamma t; RORgammaT; Rorc2; Rorg; TOR; Thor; Nr1f3
Host Species Mouse
Immunogen Mouse RORγt Recombinant Protein
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19885
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.