Learn More
ACROBiosystems Recombinant Protein;Human ROR2 / NTRKR2 Protein, Fc Tag;HEK293;50UG;RO2,ROR2,NTRKR2,ROR-2,NTRKR-2
Supplier:  ACROBiosystems RO2H525150UG
Encompass_Preferred
Recombinant Protein;Human ROR2 / NTRKR2 Protein, Fc Tag;50UG;Human ROR2, Fc Tag (RO2-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # AAI30523.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8628
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.