Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RORB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530152 Supplier Invitrogen™ Supplier No. PA530152
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, mouse brain. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (86%), Chicken (96%), Sheep (96%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RORB
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene RORB
Gene Accession No. P45446, Q8R1B8, Q92753
Gene Alias bA133M9.1; Nr1f2; Nuclear receptor ROR-beta; nuclear receptor RZR-beta; Nuclear receptor subfamily 1 group F member 2; RAR related orphan receptor B; RAR-related orphan receptor B; RAR-related orphan receptor beta; retinoic acid-binding receptor beta; retinoid-related orphan receptor b; retinoid-related orphan receptor beta; retinoid-related orphan receptor-beta; ror beta; Rorb; Rorbeta; ROR-BETA; rzrb; RZR-beta; sb:eu943; transcription factor; Unknown (protein for IMAGE:7173671)
Gene Symbols RORB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ROR beta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225998, 309288, 6096
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.