Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RORC Monoclonal Antibody (OTI3C9), TrueMAB™, OriGene
SDP

Catalog No. p-200014223
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2891 Supplier OriGene Supplier No. TA809572
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RORC (RAR related orphan receptor C) is a DNA-binding transcription factor and is a member of the NR1 subfamily of nuclear hormone receptors. The specific functions of RORC are not known; however, studies of a similar gene in mice have shown that this gene may be essential for lymphoid organogenesis and may play an important regulatory role in thymopoiesis. In addition, studies in mice suggest that the protein encoded by the RORC gene may inhibit the expression of Fas ligand and IL2. Diseases associated with RORC include Immunodeficiency 42 and Anus Cancer.

Specifications

Antigen RORC
Applications Western Blot
Classification Monoclonal
Clone OTI3C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RORC
Gene Accession No. P51449
Gene Alias IMD42; NR1F3; RORG; RZR-GAMMA; RZRG; TOR; RORgt
Gene Symbols RORC
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-266 of human RORC produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6097
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.