Learn More
Cayman Chemical ROS Detection Cell-Based Assay Kit (DHE), 96 wells
SDP
Supplier:  Cayman Chemical 60129096WELLS
Encompass_Preferred
Dihydroethidium (hydroethidine or DHE) is a widely used ethidium-based, redox-sensitive fluorescent probe. DHE has been shown to be oxidized by superoxide to form 2-hydroxyethidium (2-OH-E+) (ex 500-530 nm/em 590-620 nm) or by non-specific oxidation by other sources of ROS to form ethidium (E+) (ex 480 nm/em 576 nm).{32281,32282} Given their narrow spectral range, distinguishing between the two species using filter-based optical systems is often difficult and can lead to misreporting of the species of ROS being generated. This assay kit uses DHE as a fluorescent probe for the detection of ROS generation. Antimycin A, an inhibitor of complex III of the mitochondrial electron transport chain, is included as a positive control for ROS generation. N-acetyl Cysteine is included as an antioxidant control.
Catalog No. NC1310639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.