Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roseoflavin, ≥97%, MP Biomedicals™

Catalog No. ICN15425225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
0.1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15425225 25 mg
ICN15425210 10 mg
ICN15425280 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15425225 Supplier MP Biomedicals, Inc Supplier No. 0215425225
Add to Cart
Add to Cart
Antibacterial
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51093-55-1
Molecular Formula C18H23N5O6
Molecular Weight (g/mol) 405.411
InChI Key IGQLDUYTWDABFK-GUTXKFCHSA-N
Synonym roseoflavin, roseoflavine, 8-demethyl-8-dimethylamino riboflavin, 1-deoxy-1-8-dimethylamino-7-methyl-2,4-dioxo-3,4-dihydrobenzo g pteridin-10 2h-yl-d-ribitol
PubChem CID 49867612
ChEBI CHEBI:72346
IUPAC Name 8-(dimethylamino)-7-methyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
SMILES CC1=CC2=C(C=C1N(C)C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(CO)O)O)O

Specifications

Color Brown
Assay Percent Range ≥97%
Quantity 25 mg
Formula Weight 405.4
Physical Form Solid
Chemical Name or Material Roseoflavin
Recommended Storage Store at 0°C, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.