Learn More
Cayman Chemical Rosiglitazone, 122320-73-4, 100 mg
SDP
Supplier:  Cayman Chemical 71740100
Encompass_Preferred
A thazolidinedione agonist of PPARγ; binds to the LBD of PPARγ (Kd = 43 nM); selectively activates chimeras containing the LBDs of PPARγ over PPARα and PPARδ in a cell-based reporter assay at 10 μM; activates full-length PPARγ1 and PPARγ2 in a reporter assay (EC50s = 30 and 100 nM, respectively); induces differentiation of C3H10T1/2 stem cells to adipocytes at 1 μM; decreases HbA1c and fasting blood glucose levels in a rat model of type 2 diabetes induced by STZ and a high-carbohydrate/high-fat diet at 4 mg/kg; inhibits increases in contusion volume, macrophage infiltration and activation of microglia, and expression of IL-6, MCP1, ICAM1, caspase-3, and Bax in mouse cerebral cortex in a model of traumatic brain injury induced by controlled cortical impact at 6 mg/kg
Catalog No. NC9560589
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.