Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Rosiglitazone

Click to view available options
Quantity:
10 mg
50 mg
Description
Rosiglitazone is a potent and selective PPARγ agonist (EC50 = 60 nM); exhibits no activity at PPARα and PPARβ. Promotes differentiation of pluripotent C3H10T1/2 stem cells into adipocytes. Also promotes differentiation of urothelial organoids in combination with Erlotinib (Cat. No. 7194). Exhibits antihyperglycemic activity in diabetic ob/ob mouse model. Antidiabetic agent.
Specifications
Specifications
| Chemical Name or Material | 5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione |
| CAS | 122320-73-4 |
| Synonym | BRL 49653 |
| Quantity | 10 mg |
| Target | PPAR gamma Receptor Agonists |
| Molecular Formula | C18H19N3O3S |
| Formula Weight | 357.43 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction