Learn More
Enzo Life Sciences Rosiglitazone, (25mg), CAS Number: 122320-73-4
SDP
Supplier:  Enzo Life Sciences ALX350125M025
Encompass_Preferred
Insulin sensitizing agent. Potent insulin sensitizing agent. Peroxisome proliferator-activated receptor γ (PPAR-γ) agonist. Hepatotoxic. Antidiabetic. Formula: C18H19N3O3S. MW: 357.4. CAS Number: 122320-73-4. MI: 14: 8265. Purity: ≥97% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (>25mg/ml), 100% ethanol or methanol. Use/Stability: Stable for 2 years after receipt when stored at +4°C. Stock solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: +4°C
Catalog No. NC9331406
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.