Learn More
Abcam Rosiglitazone Potassium Salt, PPARγ Agonist, Purity >99%, Solid, 10 mg
Description
- Molecular Weight: 395.5 Da
- Purity: >99%
- Potency: Potent, selective PPARγ agonist (Kd = 43 nM, PPARγ ligand binding domain)
- Activation: Activates PPARγ expression (EC50 = 1 μM)
- Biological Activity: Shows antidiabetic and hypoglycemic effects in vivo
- Administration: Orally active
- Solubility: Water soluble
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% Rosiglitazone potassium salt |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | PPARγ |
| Formulation | 100% Rosiglitazone potassium salt |
| Quantity | 10 mg |
| For Use With (Application) | PPARγ activation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.