Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rosiglitazone Potassium Salt, PPARgamma Agonist, Solid, >99% Purity, 50 mg

Catalog No. 502705062
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5062 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5062 Supplier Abcam Supplier No. AB14586550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent and selective PPARγ agonist with >99% purity, demonstrating antidiabetic and hypoglycemic effects.

  • Molecular weight: 395.5 Da
  • Purity: >99%
  • Potency: Potent, selective PPARγ agonist (Kd = 43 nM, PPARγ ligand binding domain)
  • Activation: Activates PPARγ expression (EC50 = 1 μM)
  • Biological effects: Shows antidiabetic and hypoglycemic effects in vivo
  • Properties: Orally active and water soluble
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule PPARγ
Form Solid
Content And Storage Store at -20°C.
Shipping Condition Ambient
Quantity 50 mg
For Use With (Application) PPARγ receptor activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.