Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rosin Solvent (IPA/Toluene, 50/50 v/v), Reagents
Rosin Solvent (IPA/Toluene, 50/50 v/v), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$151.60 - $690.50
Specifications
Chemical Name or Material | Rosin Solvent (IPA/Toluene, 50/50 v/v) |
---|---|
Color | Colorless |
CAS | 108-88-3, 67-63-0 |
Boiling Point | 90°C |
Grade | Reagent |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
01-336-395
|
Reagents Holdings Llc
C729705020E6 |
20 L | Steel Pail |
Each for $690.50
|
|
||||
01-336-394
|
Reagents Holdings Llc
C72970504C |
4 L | Glass Bottle |
Each for $183.90
|
|
||||
01-336-393
|
Reagents Holdings Llc
C72970504E4 |
4 L | Tin Can |
Each for $151.60
|
|
||||
Specifications
Rosin Solvent (IPA/Toluene, 50/50 v/v) | |
108-88-3, 67-63-0 | |
Reagent | |
Non-sterile |
Colorless | |
90°C | |
0.8234 | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title