Learn More
Enzo Life Sciences Rosmarinic acid (50mg). CAS: 537-15-5, 20283-92-5
SDP
Supplier:  Enzo Life Sciences ALX270253M050
Encompass_Preferred
Naturally occurring polyphenolic compound with antioxidant and anti-inflammatory properties.Inhibits several complement-dependent inflammatory processes via inhibition of the C5 convertase. Anticarcinogenic. Inhibitor of lipid peroxidation, TCR-induced T cell activation and proliferation. Alternative name: (R)-alpha-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2E-propenyl]oxy]-3,4-dihydroxy-benzenepropanoic acid. Formula: C18H16O8. MW: 360.3. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in 100% ethanol, DMSO or dimethyl formamide; also soluble in PBS. Long Term Storage: Ambient.
Catalog No. 50-200-8448
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.