Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rosmarinic Acid, 98.6%, MP Biomedicals™

Catalog No. ICN15979205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15979205 5 mg
ICN15979210 10 mg
ICN15979250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15979205 Supplier MP Biomedicals, Inc Supplier No. 0215979205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20283-92-5
Molecular Formula C18H16O8
Molecular Weight (g/mol) 360.318
MDL Number MFCD00017740
InChI Key DOUMFZQKYFQNTF-WUTVXBCWSA-N
Synonym rosmarinic acid, rosemary acid, r-rosmarinic acid, unii-mqe6xg29yi, rosmarinate, mqe6xg29yi, labiatenic acid, r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid, 3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester, 2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid
PubChem CID 5281792
ChEBI CHEBI:50371
IUPAC Name (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
SMILES C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O

Specifications

Color Beige
Assay Percent Range 0.986
Quantity 5 mg
Formula Weight 360.32
Physical Form Powder
Chemical Name or Material Rosmarinic acid, Molecular Biology
Recommended Storage Store at room temperature, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.