Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rosolic acid

Catalog No. aab2173714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2173714 25 g
AAB2173722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2173714 Supplier Thermo Scientific Chemicals Supplier No. B2173714
Only null left
Add to Cart

CAS: 603-45-2 | C19H14O3 | 290.32 g/mol

Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

Solubility
Soluble in alcohol, sodium hydroxide, potassium hydroxide and strong acids. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-45-2
Molecular Formula C19H14O3
Molecular Weight (g/mol) 290.32
MDL Number MFCD00001624
InChI Key FYEHYMARPSSOBO-UHFFFAOYSA-N
Synonym aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
PubChem CID 5100
ChEBI CHEBI:34544
IUPAC Name 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1

Specifications

Quantity 25 g
Beilstein 2055205
Merck Index 14,881
Solubility Information Soluble in alcohol,sodium hydroxide,potassium hydroxide and strong acids. Insoluble in water.
Formula Weight 290.32
Chemical Name or Material Rosolic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-041-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.