Description
- High chemical resistance
- Manufactured from a PTFE compound
- With a built-in stainless steel spring
Specifications
Specifications
| Description | Lip seal |
| Material | PTFE |
| Type | Rotary Evaporators Lip-seal |
| For Use With (Equipment) | RV 10.8001 rotary evaporators |
| Includes | Stainless Steel Spring |
| Quantity | 1 Pack |