Learn More
Vernier™ Rotary Motion Motor Kit
SDPSCI_ED
Catalog No. S16006ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Go Direct Rotary Motion Sensor
Catalog No. For Use With (Equipment)
S16006ND Go Direct Rotary Motion Sensor
1 options
Catalog No. S16006ND Supplier Vernier™ Supplier No. MKRMV
Add to Cart
Add to Cart
Includes
Rotary Motion Motor Kit
This kit includes a small electric motor with pulley, rubber band belt, motor clip, and mounting screw. The motor can be attached to the Rotary Motion Sensor, and be used as an analog tachometer, or generator. You can also perform experiments that investigate motor efficiency in different conditions.
  • This kit includes a small electric motor with pulley, rubber band belt, motor clip, and mounting screw. The motor can be attached to the Rotary Motion Sensor, and be used as an analog tachometer, or generator. You can also perform experiments that investigate motor efficiency in different conditions.
Warranty and Services
5-year limited warranty
Specifications
Includes Small electric motor with pulley, rubber band belt, motor clip, and mounting screw
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Go Direct Rotary Motion Sensor