Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Rotary Motion Sensor
SDPSCI_ED

Catalog No. S16245ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
High School and College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16245ND High School and College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16245ND Supplier Vernier™ Supplier No. RMVBTD
Add to Cart
Add to Cart

Includes

Rotary Motion Sensor, Thumb screw, 3-step pulley and mounting screw, O-ring

Requires

Requires an interface and software, sold separately

The Vernier Rotary Motion Sensor is a bidirectional angle sensor designed to measure rotational or linear position, velocity and acceleration.

Resolution: 1° or 0.25° Optical Encoder: Bidirectional, quadrature encoder Maximum Speed: 30 rev/s at 1° resolution 7.5 rev/s at 0.25° resolution 3-step Pulley: 10 mm, 29 mm and 48 mm groove diameter

Warranty and Services

5-year limited warranty

Specifications

Includes Sensor, thumb screw, 3-step pulley, mounting screws, o-ring
Grade High School and College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 1° or 0.25°
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.