Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Rotating Platform
SDPSCI_ED

Catalog No. S27842 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Anti-Slip Patches (6), Activity Guide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27842 Anti-Slip Patches (6), Activity Guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27842 Supplier United Scientific Supplies Supplier No. RTP001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For individual use to demonstrate angular momentum and centripetal force

  • Essential accessory for Bicycle Wheel Gyroscope (BWG001)
  • Use alone to demonstrate angular momentum and centripetal force
  • Plate spins freely on heavy-duty ball bearing axle
  • Features six anti-slip patches for safe classroom use
  • Activity guide included

Specifications

Product Type Rotating Platform
For Use With (Application) Demonstrating Angular Momentum and Centripetal Force
Includes Anti-Slip Patches (6), Activity Guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.