Learn More
BUCHI Rotavapor R-300 Vertical Condenser Glass Assembly
SDP
Catalog No. 05405066 Shop All BUCHI Products
Change view
Click to view available options
Material (Coating):
No Safety Coating
P+G Safety Coating
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Coating)
05-405-066 No Safety Coating
05-405-067 P+G Safety Coating
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-066 Supplier BUCHI Supplier No. 11062432
Add to Cart
Add to Cart
Vertical Condenser Glass Assembly for Rotavapor R-300, 1500 cm2. Assembly features a large top inlet for easy cleaning, connection for vapor temperature sensor, and inner glass tubing/plate. Includes a 1 liter receiving flask, tubing, and a ball joint clamp.
  • Includes receiving flask (1 L), tubing, and a ball joint clamp
  • Does NOT include evaporating flask, vacuum seal, vapor duct, and condenser holder
  • For use with Rotavapor R-300, R-2XX
Specifications
Material Borosilicate Glass (3.3)
Type Vertical Condensor
For Use With (Equipment) Rotavap™ R-300, R-210, and R-215
Includes Receiving Flask, Tubing, and a Ball Joint Clamp
Material (Coating) No Safety Coating
Volume (Metric) 1 L