Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rotavirus Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA185845 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA185845 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA185845 Supplier Invitrogen™ Supplier No. PA185845
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

PA1-85845 detects human rotavirus serotypes I, II, III, IV, and VIII. Sheep anti Rotavirus antibody recognizes human rotavirus serotypes I, II, III, IV and VIII.

Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rotavirus
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias Reoviridae
Host Species Sheep
Immunogen Five serotypes of group A Rotavirus strains Wa (serotype I), DS-1 (serotype II), Ito (III), HOCHI (IV) and 69M (VIII).
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.