Learn More
Sigma Aldrich Fine Chemicals Biosciences Rotenone, 83-79-4, MFCD09025614, 1 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences R88751G
Encompass
Empirical Formula (Hill Notation): C23H22O6, Molecular Weight: 394.42, ≥95%, bp: 210-220 °C, mp: 159-164 °C.
Catalog No. NC0684401
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.