Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rotenone
GREENER_CHOICESDP

Catalog No. 361650
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 36-165-0 Supplier Tocris Bioscience™ Supplier No. 3616/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits complex I of the mitochondrial electron transport chain

Mitochondrial electron transport chain inhibitor (IC50 = 1.7 - 2.2 μM at complex I). Inhibits NADH oxidation by cardiac sarcoplasmic reticulum (IC50 = 3.4 nM). Commonly used pesticide and induces Parkinsonism in animal models. May also inhibit autophagy induction; blocks lysosomal degradation of autophagic vacuoles. Cell-permeable and brain penetrant.

Chemical Identifiers

CAS 83-79-4
Molecular Formula C23H22O6

Specifications

Quantity 50 mg
Formula Weight 394.42
Purity >95%
Molecular Weight (g/mol) 394.423
Chemical Name or Material Rotenone
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.