Learn More
Selleck Chemical LLC Rotigotine 10mg 99755-59-6
SDP
Supplier:  Selleck Chemical LLC S427410MG
Encompass_Preferred
Rotigotine is a dopamine receptor agonist, used in the treatment of Parkinson's disease and restless legs syndrome. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3830
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.