Learn More
Scientific Industries ROTO-SHAKE GENIE 120V
Supplier:  Scientific Industries SI1100
Encompass_Preferred
Ideal for rocking, rotating, and orbital shaking. Features a stainless steel Magnetic Platform. Supplied with 1 Clip Plate each for 10-13mm Tubes, 15-17mm Tubes, and 28-30mm Tubes, 4 Magnetic Bag Mounting Strips and Non-skid Tray.
Catalog No. 50-728-088
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.