Learn More
Eppendorf™ Rotor Adapters for Centrifuge 5427 R
Catalog No. 05414335 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
1 Cryotube
1 x 1.5/2 mL Vessel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
05-414-335 1 x 1.5/2 mL Vessel
05-414-336 1 Cryotube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-414-335 Supplier Eppendorf™ Supplier No. 5820768002
Add to Cart
Add to Cart
This item is not returnable. View return policy
For 1.5/2mL tubes and cryotubes
  • Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type Microtube
Capacity 1 x 1.5/2 mL Vessel
For Use With (Equipment) Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
Tube Size 1.5/2 mL
No. per Pack Set of 4