Learn More
Eppendorf™ Rotor for Centrifuge 5420
Catalog No. 05413930 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Centrifuge 5421
Catalog No. For Use With (Equipment)
05-413-930 Centrifuge 5421
1 options
Catalog No. 05-413-930 Supplier Eppendorf™ Supplier No. 5495404007
Add to Cart
Add to Cart
Eppendorf Rotors for Centrifuge 5420, precision for diverse applications. Choose from various capacities and speeds to tailor your centrifuge setup. Accessorize with adaptors and buckets, ensuring optimal sample processing in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
TRUSTED_SUSTAINABILITY
Specifications
Description F-32x0.2-PCR - Includes rotor lid
Material Anodized Aluminum
Type Rotor
Capacity 4 x 8 PCR Tube Strips or 32 x PCR Tubes (0.2 mL)
Max. RCF 18,257 x g
Max. Speed 15,060 rpm
For Use With (Equipment) Centrifuge 5421
Includes Rotor Lid