Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Rotor F-32x0.2-PCR, for Centrifuge 5425 PROMO

Catalog No. 14285567PM Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
32 x 0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14-285-567PM 32 x 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-285-567PM Supplier Eppendorf™ Supplier No. 2231000579

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Fixed-angle rotor, for 96 x 0.2mL PCR tubes or 12 x 8-well PCR strips, includes lid

Warranty and Services

1 year

TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 x 0.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.